ContactUsLinkComponent
Aspiration station Y-shaped tubing connector
Aspiration station Y-shaped tubing connector
Catalog # GILSF1077418
Supplier:  GILSON
undefined
Aspiration station Y-shaped tubing connector
Catalog # GILSF1077418
Supplier:  GILSON
Specifications
  • Description:
    Y-tubing connector
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
The Gilson safe aspiration station Y-shaped tubing connector is for use with the Gilson safe aspiration station.
  • Connect two handles to Gilson Safe Sapiration Station station
The connector enables you to connect two handles to the station, allowing two people to use the station simultaneously.