Order Entry
Belgium
ContactUsLinkComponent
Chemicon® ReBlot Plus Mild Antibody Stripping Solution, 10X
Chemicon® ReBlot Plus Mild Antibody Stripping Solution, 10X
Catalog # MILF2502
Supplier:  Merck
CAS Number:  
undefined
Chemicon® ReBlot Plus Mild Antibody Stripping Solution, 10X
Catalog # MILF2502
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Description:
    Reblot plus mild antibody stripping solution, 10X
  • Storage conditions:
    The mild antibody stripping solution should be stored at 2...8 °C upon arrival. Product is stable for 3 - 6 months after receipt. If antibody stripping solution crystallises upon storage, it may be re-dissolved with gentle warming at 37 °C before use.
  • Application:
    Western blotting

Specifications

About this item

Western blotting is a commonly used technique for studying protein function and localisation. Typically, protein samples are electro-phoresed on SDS-page and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid based technologies, which allow reuse of southern and northern blots, it has been difficult to reuse western blots.

  • Detection method: Chemiluminescence
  • Mild antibody stripping solution (10X) - (2 containers, 25 ml each)

The MILLIPORE re-blot plus western blot recycling kit is effective for removal of antibodies from western blots that have been developed with chemiluminescence or radioactive iodine or other isotopes. It is not recommended for stripping colorimetric substrates (TMB, DAB, 4-chloronapthol, and more), as it is not possible to effectively remove substrates that precipitate at the reaction site.

The re-blot western blot recycling kit should be used only for qualitative purposes until it has been established by comparative blot analysis that stripping does not quantitatively affect a given antigen.

Caution: Note: To prevent reagent degradation secure the cap tightly upon storage. Avoid extended exposure to air.