ContactUsLinkComponent
Chemicon® ReBlot Plus Kits
Chemicon® ReBlot Plus Kits
Supplier:  Merck
Certificates
undefined
Chemicon® ReBlot Plus Kits
Supplier:  Merck
Specifications
  • Storage conditions:
    Kit components should be stored at 4 °C upon arrival. Product is stable for 3 - 6 months after receipt. If antibody stripping solution crystallises upon storage, it may be re-dissolved with gentle warming at 37 °C before use.