Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter
Catalog # COLE50109-85
Supplier: Cole-Parmer
New Product
Specifications
- Connection Type:Female Luer (port 1)
Hose barb ¹/₈" (port 2) - Flow pattern:Straight
- Description:Luer to hose barb fitting, straight adapter
- Material:PP
- Pk:10
Specifications
About this item
Cleanroom female Luer to hose barb fittings, straight adapter, ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of system.
- Animal derivative free polypropylene
- PVC-free ideal for use in bioprocess, life science, and pharmaceutical industries
- Forms a leak-free luer connection with a quick simple turn
- Single barb design tightly 'grips' the tubing
Manufactured in an ISO Class 7 certified cleanroom to minimise contamination by pollutants.
Certifications: Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant.