
Orthophosphoric acid 10% USP-NF, BP, Ph. Eur., pure, Pharma Grade
APPC147143.0716
New Product
:
- Pk:25 L
- Seq:5327
- Assay (Acidim.) (w/w) (Ph. Eur.):9.5 - 10.5 %
- Assay (Acidim.) (w/v) (USP-NF):9.5 - 10.5 %
- Identity according to Pharmacopoeias:passes test
- Appearance of solution:passes test Chloride (Cl): 0.0006 %
- Sulfate (SO4) (Ph. Eur.):0.0010 %
- Sulfate (SO4) (USP-NF):passes test
- Precipitated substances by NH4OH:passes test
- Residual solvents (Ph.Eur/USP):passes test
- Nitrate (NO3):passes test
- Hypophosphorous and phosphorous acid:passes test
- Alkaline phosphates: passes test Fe:0.0006 %
:
:98 g/mol
:1.05 g/cm³
:231-633-2
:1805
:8,III
:GHS05
:7664-38-2
:H₃PO₄
:MFCD00011340