Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nitrite Standard Solution

Nitrite Standard Solution

Nitrite standard solution, 5,0 mg/L, 1,52 mg/L NO₂-N, Plastic bottle, 500 mL
Catalog # HACHLCW960
Supplier:   Hach
Nitrite Standard Solution
Catalog # HACHLCW960
Supplier:  Hach
CAS Number:  
Nitrite Standard Solution

Nitrite standard solution, 5,0 mg/L, 1,52 mg/L NO₂-N, Plastic bottle, 500 mL

CAS Number:  
Price
71,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    5,0 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Nitrite standard solution
  • Contents:
    1,52 mg/L NO₂-N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.