Order Entry
Belgium
ContactUsLinkComponent
r-MicronSolo Wipe Rolls
r-MicronSolo Wipe Rolls
Catalog # VILE170352
Supplier:  FHP VILEDA SCS
r-MicronSolo Wipe Rolls
Catalog # VILE170352
Supplier:  FHP VILEDA SCS

Specifications

  • Packed:
    4 pack/case
  • Colour:
    Blue
  • Description:
    r-MicronSolo roll
  • L×W:
    250×320 mm
  • Seq:
    01
  • Pk:
    4 Roll

Specifications

About this item

r-MicronSolo Roll is micronfibre wipe roll made of 35% recycled fibres of post-consumer plastic. Due to its practical roll form that can be used best together with roll dispenser, easy usage and practical distribution of the wipes are guaranteed – ideal for professional cleaner's daily work.

  • Maximum hygiene due to single-use
  • Cloth is made from 35% recycled post-consumer plastics
  • Practical distribution of wipes
  • Superior micronfibre technology for advanced cleaning performance
  • High tear-resistance
  • Efficient removal of fatty dirt
  • Colour coding system

Made from the finest micronfibre, it offers an efficient and quick streak free cleaning and even removes stubborn, fatty dirt like grease and fingerprints. Due to the efficient removal of up to 99,99% of bacteria and viruses it is especially suitable for healthcare areas, where a high level of hygiene and cleanliness are required. The highly absorbent viscose fibres allow an excellent absorbency to clean big spills in just a single wipe. The new recycled wipe Roll is perfectly suitable for health care applications as well as for general building cleaning or hotels.

The advanced micronfibre technology promises rapid and effective cleaning of high-risk areas. Even delicate materials such as glass or ceramic can be cleaned with r-MicronSolo Roll without leaving any damage or marks. Due to the excellent absorbency of the r-MicronSolo Roll cleaning cloth, big spills and stubborn dirt like grease are cleaned with ease.

Keep palletised products in a cold and dry place. Avoid exposure to direct sunlight. Product is not subject to transportation regulations for hazardous substances or chemicals. Disposal via landfill or incineration possible.

This product is compliant to our internal quality standards and external regulations.