Order Entry
Belgium
ContactUsLinkComponent
Certipur® ICP Multi Element Standard Solution VI
Certipur® ICP Multi Element Standard Solution VI
Catalog # 1044880100.
Supplier:  Merck
New Product
CAS Number:  
Certipur® ICP Multi Element Standard Solution VI
Catalog # 1044880100.
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    ICP multi-element standard solution VI 30 elements in dilute nitric acid
  • Hazard class description:
    Corrosive
  • UN No.:
    2031
  • GHS signal:
    Danger
  • Storage temperature:
    15...25 °C

Specifications

About this item

Certifications: CRM certified.