Orders LinkContactUsLinkComponent
Centrifuge columns for affinity chromatography
Centrifuge columns for affinity chromatography
Centrifuge columns for affinity chromatography
Certificates
Centrifuge columns for affinity chromatography
Centrifuge columns for affinity chromatography
6 Options Available Below
Product Details & Documents
PP column, PE filter, ~30 µm pore size. Pierce™ centrifuge columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard centrifuge tubes for use in any centrifuge (1,5, 2, 15 and 50 ml).
Pierce centrifuge columns are convenient tools for handling 40 µl to 10 ml of an affinity support for purification. They can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired for binding steps with dilute samples, Column extenders can be used to increase the column reservoir capacity.
Product Family Options
Product Information
  • Resin volume
    Total volume
    Packed
    Availability
    Price
  • PIER89868
    Centrifuge columns, 0,8 ml, receiver tube fits standard microcentrifuge tubes
    Centrifuge columns, 0,8 ml, receiver tube fits standard microcentrifuge tubes
    40 - 400 µl
    800 µl
    50 columns, 50 screw caps
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 130,00 €
    130,00 €
    1 * 50 ST
       130,00 €
  • PIER89869
    Centrifuge columns, 0,8 ml, receiver tube fits standard microcentrifuge tubes
    Centrifuge columns, 0,8 ml, receiver tube fits standard microcentrifuge tubes
    40 - 400 µl
    800 µl
    4×50 columns, 4×50 screw caps
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 378,00 €
    378,00 €
    1 * 200 ST
       378,00 €
  • PIER89896
    Centrifuge columns, 2 ml, receiver tube fits standard 15 ml conical centrifuge tubes
    Centrifuge columns, 2 ml, receiver tube fits standard 15 ml conical centrifuge tubes
    2 ml
    5 ml (2 ml resiiniä, 3 ml vapaatilavuus)
    25 columns, 25 screw caps and tips
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 80,00 €
    80,00 €
    1 * 25 ST
       80,00 €
  • PIER89897
    Centrifuge columns, 5 ml, receiver tube fits standard 15 ml conical centrifuge tubes
    Centrifuge columns, 5 ml, receiver tube fits standard 15 ml conical centrifuge tubes
    5 ml
    8 ml (5 ml resiiniä, 3 ml vapaatilavuus)
    25 columns, 25 screw caps and tips
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 88,50 €
    88,50 €
    1 * 25 ST
       88,50 €
  • PIER89898
    Centrifuge columns, 10 ml, receiver tube fits standard 50 ml conical centrifuge tubes
    Centrifuge columns, 10 ml, receiver tube fits standard 50 ml conical centrifuge tubes
    10 ml
    22 ml (10 ml resiiniä, 12 ml vapaatilavuus)
    25 columns, 25 screw caps and tips
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
Accessories
Product Information
  • Resin volume
    Total volume
    Packed
    Availability
    Price
  • PIER69707
    Column extenders, PP, fits 2, 5 and 10 ml centrifuge columns
    Column extenders, PP, fits 2, 5 and 10 ml centrifuge columns
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,50 €
    52,50 €
    1 * 10 ST
       52,50 €