Order Entry
Belgium
ContactUsLinkComponent
VWR® Fixing Solution
VWR® Fixing Solution
Catalog # 87970.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Fixing Solution
Catalog # 87970.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Fixing solution

Specifications

About this item

Fixing solution with 0,02% formaldehyde in methanol 50% (v/v)).

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.