


Sodium hydroxide 0.001 mol/l (0.001 N) in methanol 50% Reag. Ph. Eur. 1081403
Catalog # 87935.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:3286
ADR:3 (6.1, 8),II
:GHS02
:GHS06
:GHS08
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548