Dichloromethane in hydrochloric acid 1% Reag. Ph. Eur. 1055901
Catalog # 87871.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: Methylene chloride
:
ADR:6.1,III
:84.93 g/mol
EINECS:200-838-9
UN:1593
:GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881