Order Entry
Belgium
ContactUsLinkComponent
Copper (II) ethylenediamine 21% in aqueous solution Reag. Ph. Eur. 3008700
Copper (II) ethylenediamine 21% in aqueous solution Reag. Ph. Eur. 3008700
Catalog # 87712.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Copper (II) ethylenediamine 21% in aqueous solution Reag. Ph. Eur. 3008700
Catalog # 87712.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :217.76 g/mol
EINECS:238-597-7
UN:1761
ADR:8 (6.1),II
 :GHS05
CAS Number:14552-35-3
 :MFCD00274628
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂