Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cell Culture Supplements and Additives
VWR® Egg Yolk Tellurite Emulsion
VWR® Egg Yolk Tellurite Emulsion
VWR® Egg Yolk Tellurite Emulsion
Catalog # 536252S
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
VWR® Egg Yolk Tellurite Emulsion
Catalog # 536252S
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Glass bottle
Pk:
50 mL
Description:
Egg yolk tellurite emulsion
Specifications
About this item
Product Details
Documentation
Product Details
Documentation