Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate 2% (w/v) in sodium hydroxide solution 0.1 mol/l Reag. Ph. Eur. 1079302
Sodium carbonate 2% (w/v) in sodium hydroxide solution 0.1 mol/l Reag. Ph. Eur. 1079302
Catalog # 87929.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium carbonate 2% (w/v) in sodium hydroxide solution 0.1 mol/l Reag. Ph. Eur. 1079302
Catalog # 87929.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494