Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1079301
Sodium carbonate 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1079301
Catalog # 87928.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium carbonate 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1079301
Catalog # 87928.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494