Order Entry
Belgium
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

Health
Supplier:   MACHEREY-NAGEL
Celite® 545, filter aid
Supplier:  MACHEREY-NAGEL
Certificates
Celite® 545, filter aid
Celite® 545, filter aid

2 Options Available Below

Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • EINECS:272-489-0
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 34,10 €
    34,10 €
    1 * 1 kg
       34,10 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 149,00 €
    149,00 €
    1 * 5 kg
       149,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.