Order Entry
Belgium
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

Health
Marketsource
Supplier:   Roth Carl
Celite® 545, filter aid
Supplier:  Roth Carl
Marketsource
Certificates
Celite® 545, filter aid

2 Options Available Below

Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • EINECS:272-489-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 24,50 €
    24,50 €
    1 * 1 kg
       24,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 45,90 €
    45,90 €
    1 * 2,5 kg
       45,90 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.