
Potassium iodide saturated solution in water Reag. Ph. Eur. 1070504
Catalog # 87911.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:166 g/mol
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405