ContactUsLinkComponent
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes
  FFCHFF5760250
 :  
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes
  FFCHFF5760250
 :  
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
 :
 :74.55 g/mol
 :1.13 g/cm³ (20 °C)
 :Ambient
 :231-211-8
 :7447-40-7
 :KCl+AgCl
 :MFCD00011360