Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium chloride
Potassium chloride 0.005 mol/l (0.005 N) in aqueous solution
Potassium chloride 0.005 mol/l (0.005 N) in aqueous solution
Potassium chloride 0.005 mol/l (0.005 N) in aqueous solution
FFCHFF21840500
:
FF CHEMICALS
:
Potassium chloride 0.005 mol/l (0.005 N) in aqueous solution
FFCHFF21840500
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
74.55 g/mol
:
Ambient
:
231-211-8
:
7447-40-7
:
KCl
:
MFCD00011360