Order Entry
Belgium
ContactUsLinkComponent
Ammonium chloride 2 mol/l (2 N) in aqueous solution
Ammonium chloride 2 mol/l (2 N) in aqueous solution
Catalog # KRAF05637.3000
Supplier:  Bernd Kraft
CAS Number:  
undefined
Ammonium chloride 2 mol/l (2 N) in aqueous solution
Catalog # KRAF05637.3000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :53.49 g/mol
Storage Temperature:Ambient
EINECS:235-186-4
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420