Order Entry
Belgium
ContactUsLinkComponent
4,5-Dibromo-2-thiophenesulphonyl chloride
4,5-Dibromo-2-thiophenesulphonyl chloride
Catalog # MAYBCD08466.0001
Supplier:  Thermo Scientific
CAS Number:  
4,5-Dibromo-2-thiophenesulphonyl chloride
Catalog # MAYBCD08466.0001
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :340.44 g/mol
Melting Pt:80 °C
UN:3261
ADR:8,II
CAS Number:81606-31-7
Formula:C₄HBr₂ClO₂S₂
MDL Number:MFCD00041439