Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromothymol blue

Bromothymol blue

Plastic bottle, 100 mL
Catalog # HACH25532
Supplier:   Hach
Bromothymol blue
Catalog # HACH25532
Supplier:  Hach
CAS Number:  76-59-5
Bromothymol blue

Plastic bottle, 100 mL

CAS Number:  76-59-5
Price
24,50 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Formula:C₂₇H₂₈Br₂O₅S
  • MDL Number:MFCD00005872
  • Molecular Weight:624.39 g/mol
  • Merck Index:13,01430
  • Melting Pt:204…202 °C
  • Storage Temperature:Ambient
  • EINECS:200-971-2

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.