Order Entry
Belgium
ContactUsLinkComponent
 
Sudan red G in toluene, Sigma-Aldrich®
 
Sudan red G in toluene, Sigma-Aldrich®
 

 

  • Appearance (colour):
    Red to dark red
  • Appearance (form):
    Powder
  • Thin layer chromatography:
    Corresponds to standard
  • Proton nmr spectrum:
    Conforms to structure
  • Wavelength (1) (uv):
    489 - 499 nm
  • Extinction (1%/1cm) (1) (uv):
    ≥ 575
  • Solvent (uv):
    Toluene

 

 

Frequently Bought Together