
1-Octanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:5324-84-5
:
Formula:C₈H₁₇NaO₃S
MDL Number:MFCD00007544
:216.28 g/mol
Melting Pt:> 300 °C
:M-Clarity™ quality level = MQ100
EINECS:226-195-4
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Hill Formula:C₈H₁₇NaO₃S
- Molar Mass:216.27 g/mol
- Melting Point:300 °C
- pH Value:5.5 - 7.5 (100 g/L, H₂O, 20 °C)
- Bulk Density:345 kg/m³
- Solubility:(20 °C) soluble
- Storage Class:10 - 13 Other liquids and solids
- WGK:WGK 2 water endangering
- R Phrase:R 36/38
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.18307.0025_TDS_EN ProductPage_118307
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_118307
SDS
Frequently Bought Together