IPTG (isopropyl-β-D-thiogalactopyranoside)
Catalog # ROCKIPTG-1000
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
:
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273