Order Entry
Belgium
ContactUsLinkComponent
CellAdhere™ Type I Collagen, Bovine, Solution
CellAdhere™ Type I Collagen, Bovine, Solution
Catalog # STMC07001
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
undefined
CellAdhere™ Type I Collagen, Bovine, Solution
Catalog # STMC07001
Fournisseur:  STEMCELL Technologies
Numéro CAS:  

Spécifications

  • Pk:
    50 mL
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    CellAdhere™ Type I collagen, Bovine, solution

Spécifications

À propos de cet article

Support cell attachment by preparing collagen-coated tissue cultureware or 3D collagen gels using CellAdhere™ Type I Collagen, Bovine, Solution. The most abundant type of collagen in the human body, type I collagen is a natural extracellular matrix protein that provides structural support and mimics physiological conditions, making it an ideal cultureware coating surface to support cell attachment.

CellAdhere™ Type I Collagen, Bovine, Solution is prepared from collagen extracted from bovine hide and has a high monomer content. The starting material is isolated from a closed herd and purified using a highly controlled manufacturing process to ensure inactivation of possible prion and/or viral contaminants. This product is sterility tested, with Endotoxin levels verified to be 0.1 EU/ml or less, and is supplied at a concentration of approximately 6 mg/ml (0,6%) aqueous solution in 0,01 M HCl (pH at about 2,0).

Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').