Order Entry
Belgium
ContactUsLinkComponent
 
Trypsin-EDTA (0,25%)
Catalog # STMC07901
CAS Number:  
undefined
Trypsin-EDTA (0,25%)
Catalog # STMC07901
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    Trypsin-EDTA (0,25%)

Specifications

About this item

Enzymatic cell dissociation reagent.

  • Dissociate cells efficiently with a combined solution
  • EDTA enhances Trypsin-mediated cell detachment

Use Trypsin-EDTA (0,25%) for enzymatic dissociation of adherent cells, cell aggregates and tissues into single-cell suspensions. EDTA weakens cell-cell adhesion, which improves trypsin's access to hydrolysis-targeted peptide bonds, and enhances detachment of adherent cells. Trypsin-EDTA is also available in a lower concentration (0,05%), for applications that require more gentle dissociation.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').