Order Entry
Belgium
ContactUsLinkComponent
Trypsin in Citrate Saline (0,25%)
Trypsin in Citrate Saline (0,25%)
Catalog # STMC07400
CAS Number:  
undefined
Trypsin in Citrate Saline (0,25%)
Catalog # STMC07400
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    Trypsin in Citrate saline (0,25%)

Specifications

About this item

0,25% porcine trypsin in citrate saline solution for the detachment of adherent cells.

  • Detach adherent cells for easy cell re-suspension and collection

Use Trypsin in Citrate saline (0,25%) for enzymatic cell dissociation. This proteolytic enzyme solution breaks down proteins to detach adherent cells in cell cultures and allows easy resuspension during cell harvesting.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').