Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

BD PrecisionGlide™ Needles

Needles, sterile, 18 G, 40 mm, Pink
  BDAM305196
 :   BD Medical
BD PrecisionGlide™ Needles
  BDAM305196
BD PrecisionGlide™ Needles
BD PrecisionGlide™ Needles

Needles, sterile, 18 G, 40 mm, Pink

 
152,00 €
   
1 * 1.000 ST
 

BD PrecisionGlide™ Needle technology incorporates a thin wall, tri-bevel tip and a proprietary lubricant which is designed to provide ease of penetration.

  • Using a BD thin-wall needle may enable you to use a smaller gauge needle as the flow rate is similar to a BD regular-wall needle one gauge larger
  • BD has 20 years experience manufacturing thin-wall needles and meet ISO standards for cannula stiffness and resistance to breakage
  • 69% of patients said they would prefer to receive a skin injection with a smaller-diameter needle

 

 
  • Colour code:
    Pink
  • Gauge:
    18 G
  • Description:
    Needles, sterile
  • Length:
    40 mm
  • Seq:
    01