CellAdhere™ Dilution Buffer
Catalog # STMC07183
Fournisseur: STEMCELL Technologies
Numéro CAS:
Spécifications
- Pk:1
- Species:Human
- Description:CellAdhere™ Dilution buffer
Spécifications
À propos de cet article
Dilution buffer for matrix proteins.
Use CellAdhere™ Dilution buffer in combination with Vitronectin XF™ cell culture matrix (developed and manufactured by Nucleus Biologics) to provide a defined, xeno-free surface for the culture of human embryonic stem (ES) and induced pluripotent stem (iPS) cells.
CellAdhere™ is required for use with Vitronectin XF™, which may be used as an alternative to Corning® Matrigel® to support the growth and differentiation of human pluripotent stem cells.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the 'Product Restrictions').