Order Entry
Belgium
ContactUsLinkComponent
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE-1/AE-3]
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE-1/AE-3]
Catalog # NOVUNBP2-29429
Supplier:  Novus Biologicals
New Product
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE-1/AE-3]
Catalog # NOVUNBP2-29429
Supplier:  Novus Biologicals
New Product

Specifications

  • Antigen name:
    Cytokeratin, pan
  • Antigen symbol:
    CK1
  • Clonality:
    Monoclonal
  • Clone:
    AE-1/AE-3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Zebrafish,
    Primates,
    Rabbit,
    Chicken,
    Reptile,
    Human,
    Bovine,
    Dog,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    KRT1
  • Antigen synonyms:
    KRT1A|NEPPK|EHK|Keratin, type II cytoskeletal 1|K1|EHK1|EPPK|AEI2
  • UniProtKB:
    Q01546, Q7Z794
  • Purity:
    Protein A or G purified
  • Storage buffer:
    10 mM PBS with 0,05% BSA, 0,05% sodium azide
  • Storage temperature:
    Store at 4 °C
  • Concentration:
    0,2 mg/ml
  • Immunogen:
    Human epidermal keratin
  • Tested applications:
    WB, flow, ICC/IF, IHC, CyTOF-ready, Dual ISH-IHC, mIF, single-cell western
  • Pk:
    0,1 mg

Specifications

About this item

Type:
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: AE-1/AE-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit, Reptile, Zebrafish