0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Tubing
Chromatography Tubing
Tubing, PEEK
Tubing, PEEK
CTSA11-1156
:
SCIENCIX
New Product
Tubing, PEEK
CTSA11-1156
:
SCIENCIX
New Product
Ext.Ø:
1,59 mm
Ø int.:
0,1778 mm
Colour:
Yellow
Length:
1,524 m
Pk:
1