Order Entry
Belgium
ContactUsLinkComponent
AmMag™ Quatro System Controllers
AmMag™ Quatro System Controllers
Catalog # GENSD00018
Supplier:  Genscript
New Product
CAS Number:  
AmMag™ Quatro System Controllers
Catalog # GENSD00018
Supplier:  Genscript
New Product
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    One controller can control up to 4 purification modules, W×D×H: 416×382×249 mm

Specifications

About this item

The AmMag Quatro is an automated solution for purification of large-scale, high quality, transfection-grade plasmids from Escherichia coli cultures.

  • Ambient temperature: 18 ~ 32 ℃
  • Relative humidity: ≤85%
  • Altitude: ≤2000 m

The solution consists of a system controller that provides simultaneous operation of up to four independent purification modules, along with pre-filled consumables and verified protocols. The consumable kit is an all-inclusive solution that contains the samples tube, reaction tube, reagent cartridge, tip box that contains the elution tube and the waste container.

Note: Avoid exposure to sources of electromagnetic interference

Sample type: Escherichia coli - E. coli
Sample throughput: Six samples per purification module, 24 samples with a four purification module setup
Plasmid sample size: Bacteria wet pellet mass ≤1,5 g
Estimated run time: ~2 hours - depending on purification program
Sample/reaction tube: AmMag Quatro 50 ml tubes
Purification method: Magnetic beads method based on alkalinelysis