Orders LinkContactUsLinkComponent
Orbipac CPS51 pH-ORP Sensor
Orbipac CPS51 pH-ORP Sensor
pH electrode including and excluding integrated NTC temperature sensor, Fibre, Handheld and laboratory measuring devices
Catalog # ENDRCPS51-1EK2FDA
Supplier:   Endress+Hauser
Orbipac CPS51 pH-ORP Sensor
Catalog # ENDRCPS51-1EK2FDA
Supplier:  Endress+Hauser
Orbipac CPS51 pH-ORP Sensor
pH electrode including and excluding integrated NTC temperature sensor, Fibre, Handheld and laboratory measuring devices
Price
345,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
pH electrode for handheld and laboratory measuring devices.
  • Fixed cable: 1,5 m
  • CPS51 excluding temperature sensor: 1EK2FDA
  • Working limit: Pressureless
Certifications: EN IEC 63000:2018.
Specifications
  • For:
    Handheld and laboratory measuring devices
  • Description:
    pH electrode including and excluding integrated NTC temperature sensor
  • Diaphr.:
    Fibre