Orders LinkContactUsLinkComponent
Label Sheets, Core2Write
Label Sheets, Core2Write
Supplier:   VELTEK ASSOCIATES
Label Sheets, Core2Write
Supplier:  VELTEK ASSOCIATES
Certificates
Label Sheets, Core2Write
Label Sheets, Core2Write
5 Options Available Below
Product Details & Documents
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
Ordering information: Also available in sheets (sold separately).
Product Family Options
Non-sterile
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • VELTC2WR2X1SHEET01
    Core2Write label, non-sterile
    Core2Write label, non-sterile
    White
    50,8×25,4 mm
    10 labels/sheet/500 sheets/case
    Log in for availability
    Log in for availability
    1 * 18.000 ST
    1 * 18.000 ST - 4 800,00 €
    4 800,00 €
    1 * 18.000 ST
       4 800,00 €
Sterile
Product Information
  • Colour
    Dimensions
    Packed
    Availability
    Price
  • VELTC2WR2X1SHEET02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    50,8×25,4 mm
    36 labels/sheet/500 sheets/case
    Log in for availability
    Log in for availability
    1 * 18.000 ST
    1 * 18.000 ST - 4 960,00 €
    4 960,00 €
    1 * 18.000 ST
       4 960,00 €
  • VELTC2WR3X1SHEET02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    76,2×25,4 mm
    18 labels/sheet, 500 sheets/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 4 800,00 €
    4 800,00 €
    1 * 10.000 ST
       4 800,00 €
  • VELTC2WR4X2SHEET02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    101,6×50,8 mm
    10 labels/sheet, 500 sheets/case
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 4 800,00 €
    4 800,00 €
    1 * 5.000 ST
       4 800,00 €
  • VELTC2WR4X3SHEET02
    Core2Write label, sterile, irradiated
    Core2Write label, sterile, irradiated
    White
    101,6×76,2 mm
    6 labels/sheet, 500 sheets/case
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 4 800,00 €
    4 800,00 €
    1 * 3.000 ST
       4 800,00 €