Order Entry
Belgium
ContactUsLinkComponent
Skeletal Muscle Cell Growth Medium
Skeletal Muscle Cell Growth Medium
Catalog # PRTV2110602
Supplier:  PROVITRO
New Product
CAS Number:  
undefined
Skeletal Muscle Cell Growth Medium
Catalog # PRTV2110602
Supplier:  PROVITRO
New Product
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    Skeletal muscle cell growth medium
  • Media Base:
    Basal Medium

Specifications

About this item

Skeletal Muscle Cell Growth Medium is a sterile liquid culture medium designed for cultivating human skeletal muscle cells (HSKMC). Delivered as a basal medium, it is suitable for culturing Provitro HSKMC upon the addition of supplement mix components.

  • Formulation is optimised for initial seeding of 4000 cells/cm², facilitating growth up to confluence (approximately 90%)
  • Medium does not require feeder layers, matrix substrates, or additional substances for effective cell cultivation

Due to the possibility of reduced proliferative activity, we recommend to use the antibiotic supplement for freshly isolated cells only. Skeletal muscle cell growth medium is thoroughly tested after each production. All components are tested in a stringent biological assay.

Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency and the population doubling time.

Storge: Store the supplemented smooth muscle cells at a temperature of −20 °C. The supplements must not be used after the expiry date. Always adhere to these storage guidelines to maintain the effectiveness and integrity.