ContactUsLinkComponent
Bi-directional/Linear shakers, Varioshake series
Bi-directional/Linear shakers, Varioshake series
Supplier:  LAUDA
Certificates
Bi-directional/Linear shakers, Varioshake series
Supplier:  LAUDA
Specifications
  • Catalog No:
  • LAUDL003058
  • LAUDL003062
  • Model:
  • VS 8 B
  • VS 15 B
  • Shaking motion:
  • Orbital
  • Orbital
  • W×D×H:
  • 350×355×160 mm
  • 480×487×160 mm
  • Weight:
  • 11 kg
  • 18 kg
  • Operating temperature range:
  • 10...50 °C
  • 10...50 °C