EasySep™ Mouse Pan-DC Enrichment Kit II
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic negative selection kit
- Fast, easy-to-use, and column-free
- Up to 65% purity
- Isolated cells are untouched
- Optimized for high yield
- Facilitates rapid flow sorting of DC subsets
The EasySep™ Mouse Pan-DC Enrichment Kit II is designed to isolate dendritic cells (DCs) from mouse splenocytes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product replaces the EasySep™ Mouse Pan-DC Enrichment Kit (Catalog #19763), for improved yield and even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together