Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Decoquinate 98%

Decoquinate 98%
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • MDL Number:MFCD00673686
  • Molecular Weight:417.55 g/mol
  • Merck Index:13,02874
  • Storage Temperature:Ambient
  • EINECS:242-389-1
Specifications for Product Family

Specification Test Results

  • Appearance:
    white to light yellow to light beige crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Melting point:
    241°C to 245°C
  • Water:
    ≤1 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 69,70 €
    69,70 €
    1 * 1 g
       69,70 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 201,00 €
    201,00 €
    1 * 5 g
       201,00 €