Order Entry
Belgium
ContactUsLinkComponent
 
Bis(4-methylphenyl)iodonium hexafluorophosphate mixture of isomers
New Product
 
Bis(4-methylphenyl)iodonium hexafluorophosphate mixture of isomers
New Product

 

  • Appearance:
    white to light yellow to light brown to light red crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Melting point:
    170°C to 180°C

 

Frequently Bought Together