Order Entry
Belgium
ContactUsLinkComponent
Ethanol ∼40%, technical (denatured with 5% methanol)
Flammable
Irritant
Health
Ethanol ∼40%, technical (denatured with 5% methanol)
Catalog # 466480050.
New Product
CAS Number:  
Flammable
Irritant
Health
Ethanol ∼40%, technical (denatured with 5% methanol)
Catalog # 466480050.
New Product
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    clear colorless liquid
  • Density:
    (20°C) 0.943 to 0.953 kg/l
  • Ethanol:
    37 to 43 v/v % (calculated)
  • Additional info:
    Denatured with 5% methanol

Specification Test Results

About this item

 :
 :46.07 g/mol
Density:0.95
Flash Pt:26°C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1987
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568