Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cryo solution, Provitro

Cryo solution, Provitro

Cryo solution, 125 ml (serum-free), 125 mL
Catalog # PRTV2040101
Supplier:   PROVITRO
Cryo solution, Provitro
Catalog # PRTV2040101
Supplier:  PROVITRO
CAS Number:  
Cryo solution, Provitro

Cryo solution, 125 ml (serum-free), 125 mL

CAS Number:  
Price
196,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

In a serum-free cell culture system, it is very important to cryopreserve cells also in a serum-free freeze medium and not in a serum containing medium. Using an ordinary serum containing freeze medium, the serum affects the cell culture system over a long period. This influence can only be removed by dilution during a large number of passages after seeding.

  • Adherent Cells: Remove adherent cells with Trypsin/EDTA. Neutralize Trypsin with Trypsin inhibito, pellet cells by centrifuging at low acceleration (200×g) for 3-5 minutes and remove Trypsin inhibitor solution. Resuspend the cells in cold Cryo-SFM at a concentration of 1 - 4 million cells/ml. Freeze the cells gradually (I °C/minute) and store them in liquid nitrogen.
  • Cells in Suspension: Pellet the cells by centrifuge at low acceleration (200×g) for 4 minutes. Remove the supernatant, gently resuspend the cells in Cryo-SFM at a concentration of 1-5 million cells/ml. Freeze the cells gradually (1 °C/minute) and store them in liquid nitrogen.
  • Thawing: Gently shake the vial in warm water (37 °C) until approximately 90% of the freeze medium is just thawed. Remove the vial immediately and continue shaking until the whole contents is thawed. (Do not allow longer incubation of the vial at 37 °C. The viability of the cells is drastically diminished in the thawed freeze medium.) Wipe it with 70% ethanol and remove the cap being careful not to touch the interior threads with fingers. Resuspend the contents of the vial gently using a pipette and dispense the contents of the vial into the equilibrated culture vessels.

Provitro's Cryo-SFM is a new formulation for cryopreservation of animal and human cells containing no serum, but rather DMSO, methyl cellulose and other cryoprotectant ingredients. After cryopreservation and thawing, a very high percentage of viable cells is obtained. Cells which are cryopreserved in Cryo-SFM also show excellent attachment ability as well as growth performance after thawing.

Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    Cryo solution, 125 ml (serum-free)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.