Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
NCC Standard Solution, 11 Components
NCC Standard Solution, 11 Components
Catalog # CMPLF115751
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
CAS Number:
Expand image
NCC Standard Solution, 11 Components
Catalog # CMPLF115751
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
CAS Number:
Specifications
Pk:
1 mL
Specifications
About this item
Product Details
Certifications:
ISO 17034, ISO 17025 and ISO 9001.
Product Details