Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium Desoxycholate

Sodium Desoxycholate

Sodium desoxycholate for bacteriological use, 500 g
Catalog # LIOF612401
Supplier:   LIOFILCHEM
Sodium Desoxycholate
Catalog # LIOF612401
Supplier:  LIOFILCHEM
CAS Number:  
Sodium Desoxycholate

Sodium desoxycholate for bacteriological use, 500 g

CAS Number:  
Price
928,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Sodium Desoxycholate is the sodium salt of desoxycholic acid, a highly purified bile acid, and inhibits Gram-positive cocci and spore forming organisms. Culture media containing sodium desoxycholate are used for the enumeration of coliforms from milk, water, sewage, etc.., and for the isolation of enteric pathogens.

  • Dehydrated powder
  • Water soluble
  • Appearance: free-flowing, homogeneous
  • Color: white
  • pH at 5% concentration: 8,5
  • Loss on drying: 2,3%

Sodium Desoxycholate can be used as an ingredient of dehydrated culture media and need dissolution in distilled or deionized water and sterilization by autoclaving.

Specifications

Specifications

  • Pk:
    500 g
  • Description:
    Sodium desoxycholate for bacteriological use
  • Cat. No.:
    LIOF612401
  • Storage:
    Store at 10...30 °C, in a dry environment

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.