Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Rouge de méthyle solution indicateur

Rouge de méthyle solution indicateur

40 mL
Catalog # ACSDCH8005022.L04
Supplier:   CPACHEM
Rouge de méthyle solution indicateur
Catalog # ACSDCH8005022.L04
Supplier:  CPACHEM
CAS Number:  493-52-7
Rouge de méthyle solution indicateur

40 mL

CAS Number:  493-52-7
Price
58,30 €
Log in for availability
1 * 40 mL
Product Details & Documents

Certifications: ISO 9001

Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specification Test Results

  • Content:
    0.1g of methyl red in 7.4ml of 0.05mol/l NaOH solution and diluted with water to 200ml.
  • Performance Test:
    passed
  • Colour change:
    from red to yellow (pH 4.2 - 6.3)

Specifications

  • Cdt:
    40 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.