Spécifications
- Media Format:Liquid
- Pk:1.000 mL
- Description:HyClone™ peak expression medium with stable L-glutamine and poloxamer 188
- Sterility:0,2 μm filtered sterile
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Phenol red:No
- Stable glutamine:Yes
- L-Glutamine:Yes
- Storage temperature:2...8 °C, away from light
- Packed:Bottle
- Shelf life:12 Months
Spécifications
À propos de cet article
Ready-to-use medium supports high cell growth, exceptional transfection efficiency, and robust viral production for adeno-associated virus and lentivirus expressed in human embryonic kidney (HEK293) cell lines.
- Ready-to-use medium contains stable L-glutamine and poloxamer 188
- Supports high transfection efficiency
- Designed for high cell yield and robust virus production
- Allows for direct adaptation
- Designed for small- to large-scale transfection and production applications
- Compatible with a variety of PEI- and lipid-based transfection reagents
HyClone™ peak expression medium is an animal-derived-component free (ADCF), hydrolysate-free and regulatory -friendly cell culture medium. The versatility of this medium allows high transfection efficiency, consistent cell growth, high viable cell density, and excellent viral titers across many HEK 293 cell lines.
This medium is designed to support outstanding performance in small- and large-scale manufacturing processes and supports seamless transition from growth to production of AAV and lentivirus using standard cell lines and processes. It is available in liquid and powder formats in user-friendly packaging.