Order Entry
Belgium
ContactUsLinkComponent
Water Detect Enterococci, Rapid Test for Enterococci in Water
Water Detect Enterococci, Rapid Test for Enterococci in Water
Catalog # LIOF630603
Supplier:  LIOFILCHEM
New Product
CAS Number:  
undefined
Water Detect Enterococci, Rapid Test for Enterococci in Water
Catalog # LIOF630603
Supplier:  LIOFILCHEM
New Product
CAS Number:  

Specifications

  • Pk:
    12
  • Description:
    Water Detect Enterococci

Specifications

About this item

Selective enrichment broth used for the presumptive identification of enterococci in the microbiological monitoring of water quality.

  • Easy to use: Transfer the content of a tube of Water Detect Enterococci in a bottle with 100 ml of the sample, shake and incubate
  • Quick results: Incubate at 35 ±2 °C for 18 to 24 h, the presence of Enterococcus spp. is indicated by a blackening of the medium
  • Storage: Room temperature (10 to 30 °C)
  • Shelf life: 2 years

Enterococci are a bacteriological indicator of fecal contamination in water. They can be included in a water monitoring program to assess quality of recreational and drinking water concerning potential risk to human health from fecal pathogens. Water Detect Enterococci allows the identification of Enterococcus species based on esculin hydrolysis.