Orders LinkContactUsLinkComponent
Stainless Chain
Stainless Chain
Stainless steel chain, 3,2 Kg, Stainless steel 316, 50 m
Catalog # SAMP5070S-50
Supplier:   Sampling Systems
Stainless Chain
Catalog # SAMP5070S-50
Supplier:  Sampling Systems
Stainless Chain
Stainless steel chain, 3,2 Kg, Stainless steel 316, 50 m
Price
738,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Our popular range of accessories are essential when sampling with Sub Samplers (Bomb Sampler) and bailers. Stainless chain provides great chemical resistance.
Specifications
  • Description:
    Stainless steel chain, 3,2 Kg
  • Length:
    50 m
  • Material:
    Stainless steel 316